Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpcat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012189.1 | NP_001012189.1 | 2 | 232 | ||
GenBank mRNA | BC089869.1 | - | 2 | 232 | 7324068 |