Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp90b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012197.2 | NP_001012197.2 | 12 | 1580 | ||
GenBank mRNA | BC081917.1 | - | 12 | 1580 | 7112411 |