Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNL4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142317.1 | NP_001135789.1 | 2 | 342 | ||
NM_001142318.1 | NP_001135790.1 | 2 | 240 | |||
NM_001324354.1 | NP_001311283.1 | 2 | 402 | |||
NM_001324355.1 | NP_001311284.1 | 2 | 402 | |||
NM_017853.2 | NP_060323.1 | 2 | 391 | |||
XM_017023377.1 | XP_016878866.1 | 2 | 407 | |||
GenBank mRNA | AK000518.1 | - | 2 | 366 | ||
AK222889.1 | - | 2 | 358 | |||
AK308891.1 | - | 2 | 308 | |||
AY566808.1 | - | 1 | 80 | |||
BC009646.1 | - | 2 | 332 | 3458545 | ||
BM849401.1 | - | 2 | 303 | |||
BP355646.1 | - | 2 | 391 | |||
CD517462.1 | - | 2 | 288 | |||
CR457240.1 | - | 1 | 80 | |||
DA499499.1 | - | 2 | 342 |