Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012213.1 | NP_001012213.1 | 11 | 1403 | ||
XM_006253645.2 | XP_006253707.1 | 11 | 1614 | |||
XM_008771502.1 | XP_008769724.1 | 11 | 1375 | |||
GenBank mRNA | BC078935.1 | - | 11 | 1403 | 7108166 | |
FQ209748.1 | - | 1356 |