Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012214.1 | NP_001012214.1 | 7 | 1068 | ||
GenBank mRNA | BC079108.1 | - | 7 | 1068 | 7125468 |