Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017864.3 | NP_060334.2 | 14, 15 | 1902 | ||
NR_073444.1 | 14, 15 | 1902 | ||||
NR_073445.1 | 14, 15 | 1871 | ||||
XM_017013616.1 | XP_016869105.1 | 14, 15 | 1902 | |||
XM_017013617.1 | XP_016869106.1 | 14, 15 | 1902 | |||
XM_017013618.1 | XP_016869107.1 | 11, 12 | 1455 | |||
XM_017013619.1 | XP_016869108.1 | 14, 15 | 1861 | |||
GenBank mRNA | AB161944.1 | - | 14, 15 | 1886 | ||
AK000537.1 | - | 4, 5 | 456 | |||
AK001403.1 | - | 7, 8 | 842 | |||
AK091278.1 | - | 13, 14 | 1639 | |||
AK225533.1 | - | 4, 5 | 456 | |||
BC050536.1 | - | 6, 7 | 690 | |||
BC064915.1 | - | 6, 7 | 731 | |||
BC136754.1 | - | 14, 15 | 1816 | 9020744 | ||
BC144396.1 | - | 14, 15 | 1816 | |||
BC144397.1 | - | 14, 15 | 1785 | |||
BX538203.1 | - | 10, 11 | 1225 |