Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136036.2 | NP_001129508.1 | 6 | 1014 | ||
NM_017865.3 | NP_060335.2 | 6 | 798 | |||
XM_011544220.1 | XP_011542522.1 | 6 | 1055 | |||
XM_011544221.1 | XP_011542523.1 | 6 | 1055 | |||
XM_011544222.1 | XP_011542524.1 | 6 | 670 | |||
XM_011544227.1 | XP_011542529.1 | 6 | 966 | |||
XM_017001698.1 | XP_016857187.1 | 6 | 670 | |||
XM_017001699.1 | XP_016857188.1 | 6 | 1055 | |||
XM_017001700.1 | XP_016857189.1 | 6 | 670 | |||
XM_017001701.1 | XP_016857190.1 | 6 | 785 | |||
XM_017001702.1 | XP_016857191.1 | 6 | 785 | |||
XM_017001703.1 | XP_016857192.1 | 6 | 985 | |||
XM_017001704.1 | XP_016857193.1 | 6 | 942 | |||
XM_017001705.1 | XP_016857194.1 | 6 | 845 | |||
GenBank mRNA | AB209706.1 | - | 3 | 763 | ||
AK298899.1 | - | 5 | 654 | |||
AK302187.1 | - | 6 | 948 | |||
AL133100.1 | - | 4 | 1527 | |||
BC002948.1 | - | 6 | 754 | 3533691 |