Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOHLH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012415.2 | NP_001012415.2 | 7 | 1211 | ||
GenBank mRNA | AY902244.1 | - | 7 | 1211 | ||
BC031861.1 | - | 7 | 1222 | 4826967 |