Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012466.1 | NP_001012484.1 | 4 | 1061 | ||
GenBank mRNA | AY568760.1 | - | 4 | 1061 |