Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012473.1 | NP_001012491.1 | 9 | 1045 | ||
GenBank mRNA | BC089905.1 | - | 9 | 1045 | 7322752 | |
FQ218345.1 | - | 9 | 963 | |||
FQ225242.1 | - | 9 | 1065 | |||
FQ226282.1 | - | 9 | 1086 | |||
FQ227427.1 | - | 9 | 1067 | |||
FQ235323.1 | - | 9 | 1084 |