Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202403.1 | NP_001189332.1 | 2 | 313 | ||
NM_017873.3 | NP_060343.1 | 2 | 313 | |||
NM_177999.2 | NP_821066.1 | 2 | 313 | |||
GenBank mRNA | AK000555.1 | - | 2 | 310 | ||
AK023004.1 | - | 2 | 259 | |||
AK225618.1 | - | 2 | 288 | |||
AK292807.1 | - | 2 | 310 | |||
BC001719.1 | - | 2 | 268 | |||
BC065913.1 | - | 2 | 216 | 5211288 | ||
BX648923.1 | - | 1 | 834 | |||
DA953275.1 | - | 2 | 313 |