Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162491.1 | NP_001155963.1 | 8 | 876 | ||
NR_103708.1 | 9 | 1057 | ||||
NR_103709.1 | 9 | 1283 | ||||
XM_011530950.2 | XP_011529252.1 | 8 | 1799 | |||
GenBank mRNA | BC140808.1 | - | 8 | 950 | 9021676 | |
BC144603.1 | - | 8 | 950 | 9053134 |