Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162491.1 | NP_001155963.1 | 4 | 287 | ||
NR_103708.1 | 4 | 287 | ||||
NR_103709.1 | 4 | 287 | ||||
XM_011530950.2 | XP_011529252.1 | 4 | 984 | |||
XM_011530951.2 | XP_011529253.1 | 4 | 984 | |||
GenBank mRNA | AK301813.1 | - | 4 | 287 | ||
BC140808.1 | - | 4 | 361 | 9021676 | ||
BC144603.1 | - | 4 | 361 | 9053134 | ||
DB727708.1 | - | 4 | 284 |