Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1c8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013067.2 | NP_001013085.1 | 2, 3 | 203 |
Gene Symbol: LOC103690048 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759444.1 | XP_008757666.1 | 2, 3 | 242 |