Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013086.1 | NP_001013104.1 | 9 | 1457 | ||
XM_006236661.2 | XP_006236723.1 | 10 | 1297 | |||
XM_006236662.2 | XP_006236724.1 | 10 | 1390 | |||
XM_006236663.2 | XP_006236725.1 | 10 | 1524 | |||
XM_006236666.2 | XP_006236728.1 | 9 | 1244 | |||
GenBank mRNA | BC079104.1 | - | 9 | 1457 | 7125038 |