Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013190.1 | NP_001013208.1 | 9 | 1367 | ||
XM_006255276.2 | XP_006255338.1 | 9 | 1842 | |||
XM_006255277.2 | XP_006255339.1 | 9 | 1839 | |||
GenBank mRNA | BC084695.1 | - | 9 | 1367 | 7114647 |