Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013190.1 | NP_001013208.1 | 8 | 1002 | ||
XM_006255276.2 | XP_006255338.1 | 8 | 1066 | |||
XM_006255277.2 | XP_006255339.1 | 8 | 1063 | |||
GenBank mRNA | BC084695.1 | - | 8 | 1002 | 7114647 |