Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013191.1 | NP_001013209.1 | 5 | 702 | ||
XM_006255502.2 | XP_006255564.1 | 5 | 696 | |||
GenBank mRNA | BC081946.1 | - | 5 | 702 | 7121452 |