Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tial1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013193.1 | NP_001013211.1 | 8 | 859 | ||
XM_006230310.2 | XP_006230372.1 | 8 | 1065 | |||
XM_006230312.2 | XP_006230374.1 | 8 | 1116 | |||
XM_006230313.2 | XP_006230375.1 | 8 | 1116 | |||
XM_008759902.1 | XP_008758124.1 | 8 | 884 | |||
GenBank mRNA | BC091409.1 | - | 8 | 859 | 7325481 |