Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013196.1 | NP_001013214.1 | 2, 3 | 233 | ||
XM_006230861.2 | XP_006230923.1 | 1, 2 | 388 | |||
XM_006230862.2 | XP_006230924.1 | 2 | 408 | |||
GenBank mRNA | BC088201.1 | - | 2, 3 | 233 | 7378025 |