Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anp32e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013200.1 | NP_001013218.1 | 2 | 428 | ||
GenBank mRNA | BC083744.1 | - | 2 | 428 | 7192147 |