Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013210.1 | NP_001013228.1 | 4 | 699 | ||
GenBank mRNA | BC083746.1 | - | 4 | 699 | 7192246 |