Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013220.2 | NP_001013238.2 | 15 | 4659 | ||
XM_006242010.2 | XP_006242072.1 | 6 | 498 | |||
XM_008765758.1 | XP_008763980.1 | 6 | 762 | |||
XM_008765759.1 | XP_008763981.1 | 6 | 762 | |||
XM_008765760.1 | XP_008763982.1 | 6 | 498 | |||
GenBank mRNA | BC088419.1 | - | 6 | 474 | ||
BC169041.1 | - | 6 | 438 | |||
EF203432.1 | - | 5 | 348 | |||
EF203433.1 | - | 5 | 348 |