Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013235.1 | NP_001013253.1 | 4 | 280 | ||
XM_006255292.2 | XP_006255354.1 | 4 | 428 | |||
XM_006255293.2 | XP_006255355.1 | 4 | 309 | |||
XM_006255295.2 | XP_006255357.1 | 4 | 307 | |||
GenBank mRNA | BC076390.1 | - | 4 | 280 | 7106066 | |
BC086417.1 | - | 4 | 280 | 7127948 |