Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013245.1 | NP_001013263.1 | 3, 4 | 269 | ||
XM_008760682.1 | XP_008758904.1 | 4, 5 | 411 | |||
GenBank mRNA | BC058155.1 | - | 3, 4 | 269 | 6888541 |