Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013246.1 | NP_001013264.1 | 5, 6 | 291 | ||
GenBank mRNA | AY935257.1 | - | 5, 6 | 291 |