Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013252.1 | NP_001013270.1 | 7 | 854 | ||
GenBank mRNA | BC098951.1 | - | 846 | 7324816 |