Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkain2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013411.2 | NP_001013429.2 | 3 | 448 | ||
NM_001025286.2 | NP_001020457.2 | 3 | 448 | |||
GenBank mRNA | AY940478.1 | - | 3 | 244 | ||
BC141063.1 | - | 3 | 318 | 9055675 | ||
DQ065606.1 | - | 3 | 381 | |||
DQ065607.1 | - | 3 | 381 | |||
EF058049.2 | - | 3 | 381 | |||
EF058050.1 | - | 2 | 229 | |||
EF058051.1 | - | 3 | 229 |