Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc6l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013608.2 | NP_001013626.2 | 19 | 4747 | ||
GenBank mRNA | AK006112.1 | - | 3 | 1161 | ||
BC026917.1 | - | 1 | 544 | |||
BC075679.1 | - | 4 | 1758 |