Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013653.2 | NP_001013675.1 | 1 | 419 | ||
GenBank mRNA | BC140911.1 | - | 1 | 388 | 9056723 | |
DN996987.1 | - | 1 | 425 | |||
DQ355157.1 | - | 1 | 355 |