Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDPGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013657.2 | NP_001013679.2 | 4 | 988 | ||
NM_001322811.1 | NP_001309740.1 | 5 | 1107 | |||
GenBank mRNA | AK130527.1 | - | 4 | 1574 |