Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021090.3 | NP_066576.1 | 20 | 7916 | ||
NM_153050.2 | NP_694690.1 | 20 | 7832 | |||
NM_153051.2 | NP_694691.1 | 19 | 7805 | |||
XM_005261803.2 | XP_005261860.1 | 22 | 8061 | |||
XM_005261804.3 | XP_005261861.1 | 21 | 7978 | |||
XM_005261805.2 | XP_005261862.1 | 21 | 8034 | |||
XM_005261806.2 | XP_005261863.1 | 21 | 7943 | |||
XM_017029028.1 | XP_016884517.1 | 20 | 7835 | |||
XM_017029029.1 | XP_016884518.1 | 19 | 7798 | |||
XM_017029030.1 | XP_016884519.1 | 19 | 7730 | |||
XM_017029031.1 | XP_016884520.1 | 18 | 7691 | |||
XM_017029033.1 | XP_016884522.1 | 15 | 7262 | |||
XM_017029034.1 | XP_016884523.1 | 21 | 9265 | |||
GenBank mRNA | AK056895.1 | - | 1 | 1559 |