Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099686.2 | NP_001093156.1 | 4 | 538 |
Gene Symbol: NXF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022053.3 | NP_071336.1 | 4 | 538 | ||
GenBank mRNA | AF285596.1 | - | 1 | 137 | ||
AJ277659.1 | - | 8 | 1024 | |||
AK308255.1 | - | 4 | 477 | |||
BC015020.1 | - | 4 | 458 | 3921074 |