Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC401357 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130727.1 | 1 | 2157 | |||
GenBank mRNA | AK125790.1 | - | 1 | 2156 | ||
AK128114.1 | - | 1 | 1898 | |||
BC137417.1 | - | 1 | 1891 |
Gene Symbol: LOC100132217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128836.1 | - | 3 | 2423 |
Gene Symbol: LOC107986805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012932.1 | XP_016868421.1 | 1 | 639 |