Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013694.2 | NP_001013716.2 | 5 | 666 | ||
XM_011530178.2 | XP_011528480.1 | 5 | 729 | |||
GenBank mRNA | AK293916.1 | - | 6 | 646 | ||
BC066962.1 | - | 6 | 649 | 4820477 |