Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145725.1 | NP_001139197.1 | 4 | 344 | ||
NM_017816.2 | NP_060286.1 | 4 | 442 | |||
XM_011513505.1 | XP_011511807.1 | 4 | 744 | |||
XM_011513506.2 | XP_011511808.1 | 3 | 375 | |||
GenBank mRNA | AK000432.1 | - | 4 | 389 | ||
AK290750.1 | - | 4 | 419 | |||
AL136750.1 | - | 4 | 344 | |||
BC015796.2 | - | 4 | 404 | 3911275 | ||
CR533548.1 | - | 2 | 201 |