Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242.4 | NP_001233.1 | 1, 2 | 355 | ||
XM_017020232.1 | XP_016875721.1 | 1, 2 | 373 | |||
XM_017020234.1 | XP_016875723.1 | 1, 2 | 373 | |||
GenBank mRNA | AK315732.1 | - | 1, 2 | 374 | ||
BC012160.1 | - | 1, 2 | 341 | 4575359 | ||
BI225100.1 | - | 1, 2 | 399 | |||
M63928.1 | - | 1, 2 | 244 |