Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iigp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013828.2 | NP_001013850.1 | 219 | |||
GenBank mRNA | BC020118.1 | - | 220 | 3675329 |