Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC088269.1 | - | 260 |
Gene Symbol: IgG-2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC088240.1 | - | 2 | 265 | 7365825 |
Gene Symbol: LOC366747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC088184.1 | - | 3 | 260 |