Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014152.1 | NP_001014174.1 | 22 | 2169 | ||
XM_006255269.2 | XP_006255331.1 | 22 | 2206 | |||
XM_006255270.2 | XP_006255332.1 | 22 | 2206 | |||
XM_008772410.1 | XP_008770632.1 | 22 | 2918 | |||
GenBank mRNA | BC085909.1 | - | 22 | 2169 | 7096506 |