Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014159.2 | NP_001014181.2 | 10 | 1470 | ||
XM_006228781.2 | XP_006228843.1 | 9 | 1408 | |||
GenBank mRNA | BC079321.1 | - | 10 | 1428 | 7135399 | |
FQ216075.1 | - | 6 | 852 | |||
FQ235070.1 | - | 6 | 820 |