Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014180.1 | NP_001014202.1 | 10 | 1237 | ||
XM_006234585.2 | XP_006234647.1 | 10 | 1315 | |||
GenBank mRNA | BC083756.1 | - | 10 | 1237 | 7192721 |