Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mall | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014182.1 | NP_001014204.1 | 4 | 1168 | ||
XM_008762181.1 | XP_008760403.1 | 4 | 708 | |||
GenBank mRNA | BC090079.1 | - | 4 | 1168 | 7126923 |