Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014184.1 | NP_001014206.1 | 13 | 1775 | ||
XM_006235487.2 | XP_006235549.1 | 15 | 3200 | |||
XM_006235489.2 | XP_006235551.1 | 15 | 2551 | |||
XM_006235490.2 | XP_006235552.1 | 16 | 3345 | |||
XM_006235491.2 | XP_006235553.1 | 15 | 2811 | |||
XM_006235493.2 | XP_006235555.1 | 14 | 2345 | |||
XM_006235494.2 | XP_006235556.1 | 14 | 2994 | |||
GenBank mRNA | BC089878.1 | - | 13 | 1775 | 7326188 |