Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myzap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014211.1 | NP_001014233.1 | 13 | 1386 | ||
XM_008766312.1 | XP_008764534.1 | 14 | 1572 | |||
GenBank mRNA | BC089899.1 | - | 13 | 1386 | 7382871 |