Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhej1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014217.1 | NP_001014239.1 | 8 | 974 | ||
XM_006245240.1 | XP_006245302.1 | 8 | 1052 | |||
XM_006245241.2 | XP_006245303.1 | 8 | 1628 | |||
XM_006245242.2 | XP_006245304.1 | 8 | 1397 | |||
GenBank mRNA | BC079033.1 | - | 8 | 974 | 7114269 |