Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014219.1 | NP_001014241.1 | 3 | 589 | ||
XM_006245248.2 | XP_006245310.1 | 3 | 642 | |||
GenBank mRNA | BC081973.1 | - | 3 | 589 | 7126276 |