Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014224.2 | NP_001014246.1 | 7 | 1261 | ||
GenBank mRNA | AB294402.1 | - | 7 | 1261 | ||
BC079040.1 | - | 7 | 1333 | 7114771 |