Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014229.1 | NP_001014251.1 | 15 | 1390 | ||
GenBank mRNA | BC085872.1 | - | 15 | 1390 | 7191363 |