Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem55b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014233.2 | NP_001014255.1 | 7 | 1543 | ||
XM_006251888.1 | XP_006251950.1 | 7 | 1516 | |||
XM_006251889.2 | XP_006251951.1 | 6 | 1402 | |||
GenBank mRNA | BC087604.1 | - | 7 | 1546 | 7312219 | |
FQ225412.1 | - | 7 | 1521 |